Anti-KLRK1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 149810]

Supplier: R & D Systems
FAB139G-025 FAB139G-100
102880-348EA 188 USD
102880-348 102880-350
Anti-KLRK1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 149810]
Antibodies
Type: Primary
Antigen: KLRK1
Clonality: Monoclonal
Clone: 149810
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR