Anti-KLRK1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 149810]

Supplier: R & D Systems
FAB139A-100 FAB139A-025
102880-342EA 543.75 USD
102880-342 102880-340
Anti-KLRK1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 149810]
Antibodies
Type: Primary
Antigen: KLRK1
Clonality: Monoclonal
Clone: 149810
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR