Anti-KLRC2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 134591]

Supplier: R & D Systems
FAB138P-100 FAB138P-025
102880-336EA 244.4 USD
102880-336 102880-338
Anti-KLRC2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 134591]
Antibodies
Type: Primary
Antigen: KLRC2
Clonality: Monoclonal
Clone: 134591
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR