Anti-KLRC2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 134591]

Supplier: R & D Systems
FAB138A-100 FAB138A-025
102881-162EA 647.88 USD
102881-162 102880-824
Anti-KLRC2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 134591]
Antibodies
Type: Primary
Antigen: KLRC2
Clonality: Monoclonal
Clone: 134591
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR