Anti-ULBP1 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 170818]

Supplier: R & D Systems
FAB1380C
102880-814EA 633.42 USD
102880-814
Anti-ULBP1 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 170818]
Antibodies
Type: Primary
Antigen: ULBP1
Clonality: Monoclonal
Clone: 170818
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR