Anti-ULBP1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 170818]

Supplier: R & D Systems
FAB1380A
102880-812EA 639.2 USD
102880-812
Anti-ULBP1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 170818]
Antibodies
Type: Primary
Antigen: ULBP1
Clonality: Monoclonal
Clone: 170818
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR