Anti-KIT Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 180627]

Supplier: R & D Systems
FAB1356A
220017-048EA 323.94 USD
220017-048
Anti-KIT Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 180627]
Antibodies
Type: Primary
Antigen: KIT
Clonality: Monoclonal
Clone: 180627
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR