Anti-ITGA4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7.2R]

Supplier: R & D Systems
FAB1354P
220017-047EA 415.05 USD
220017-047
Anti-ITGA4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7.2R]
Antibodies
Type: Primary
Antigen: ITGA4
Clonality: Monoclonal
Clone: 7.2R
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR