Anti-ITGA4 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 7.2R]

Supplier: R & D Systems
FAB1354G
102880-802EA 415.05 USD
102880-802
Anti-ITGA4 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 7.2R]
Antibodies
Type: Primary
Antigen: ITGA4
Clonality: Monoclonal
Clone: 7.2R
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR