Anti-ITGA6 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: GoH3]

Supplier: R & D Systems
FAB13501A
220017-044EA 454.09 USD
220017-044
Anti-ITGA6 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: GoH3]
Antibodies
Type: Primary
Antigen: ITGA6
Clonality: Monoclonal
Clone: GoH3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR