Anti-FCGR2A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 190723]

Supplier: R & D Systems
FAB1330P
220017-040EA 513.38 USD
220017-040
Anti-FCGR2A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 190723]
Antibodies
Type: Primary
Antigen Symbol: FCGR2A
Antigen Name: Fc fragment of IgG, low affinity IIa, receptor (CD32)
Antigen Synonyms: CD32, CD32A, CDw32, FCG2, FCGR2, FCGR2A1, FcGR, IGFR2
Clonality: Monoclonal
Clone: 190723
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR