Anti-FCGR2A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 190723]

Supplier: R & D Systems
FAB1330A
102880-796EA 527.85 USD
102880-796
Anti-FCGR2A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 190723]
Antibodies
Type: Primary
Antigen Symbol: FCGR2A
Antigen Name: Fc fragment of IgG, low affinity IIa, receptor (CD32)
Antigen Synonyms: CD32, CD32A, CDw32, FCG2, FCGR2, FCGR2A1, FcGR, IGFR2
Clonality: Monoclonal
Clone: 190723
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR