Anti-O4 Mouse Monoclonal Antibody [clone: O4] (PE (Phycoerythrin))

Supplier: R & D Systems
FAB1326P
220017-039EA 614.62 USD
220017-039
Anti-O4 Mouse Monoclonal Antibody [clone: O4] (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen Symbol:
Antigen Name: Oligodendrocyte Marker O4
Antigen Synonyms:
Clonality: Monoclonal
Clone: O4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse, Rat, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR