Anti-O4 Mouse Monoclonal Antibody [clone: O4] (Alexa Fluor® 488)

Supplier: R & D Systems
FAB1326G
102880-794EA 639.2 USD
102880-794
Anti-O4 Mouse Monoclonal Antibody [clone: O4] (Alexa Fluor® 488)
Antibodies
Type: Primary
Antigen Symbol:
Antigen Name: Oligodendrocyte Marker O4
Antigen Synonyms:
Clonality: Monoclonal
Clone: O4
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse, Rat, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR