Anti-MICA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 159227]

Supplier: R & D Systems
FAB1300P-025 FAB1300P-100
102881-156EA 244.4 USD
102881-156 102881-158
Anti-MICA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 159227]
Antibodies
Type: Primary
Antigen: MICA
Clonality: Monoclonal
Clone: 159227
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR