Anti-MICA Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 159227]

Supplier: R & D Systems
FAB1300A-025 FAB1300A-100
102881-148EA 229.94 USD
102881-148 102881-150
Anti-MICA Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 159227]
Antibodies
Type: Primary
Antigen: MICA
Clonality: Monoclonal
Clone: 159227
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR