Anti-MICA+MICB Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 159207]

Supplier: R & D Systems
FAB13001P-100 FAB13001P-025
102881-144EA 202.46 USD
102881-144 102881-146
Anti-MICA+MICB Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 159207]
Antibodies
Type: Primary
Antigen: MICA+MICB
Clonality: Monoclonal
Clone: 159207
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR