Anti-MICA+MICB Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 159207]

Supplier: R & D Systems
FAB13001N-100 FAB13001N-025
102880-324EA 284.89 USD
102880-324 102880-326
Anti-MICA+MICB Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 159207]
Antibodies
Type: Primary
Antigen: MICA+MICB
Clonality: Monoclonal
Clone: 159207
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR