Anti-MICA+MICB Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 159207]

Supplier: R & D Systems
FAB13001A-025 FAB13001A-100
102880-316EA 205.35 USD
102880-316 102880-318
Anti-MICA+MICB Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 159207]
Antibodies
Type: Primary
Antigen: MICA+MICB
Clonality: Monoclonal
Clone: 159207
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR