Anti-ULBP-2/5/6 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 165903]

Supplier: R & D Systems
FAB1298P
220017-031EA 614.62 USD
220017-031
Anti-ULBP-2/5/6 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 165903]
Antibodies
Type: Primary
Antigen: ULBP-2/5/6
Clonality: Monoclonal
Clone: 165903
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR