Anti-ULBP-2/5/6 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 165903]

Supplier: R & D Systems
FAB1298A
220017-030EA 655.11 USD
220017-030
Anti-ULBP-2/5/6 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 165903]
Antibodies
Type: Primary
Antigen: ULBP-2/5/6
Clonality: Monoclonal
Clone: 165903
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR