Anti-TREM1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 193015]

Supplier: R & D Systems
FAB1278G
102880-312EA 561.11 USD
102880-312
Anti-TREM1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 193015]
Antibodies
Type: Primary
Antigen: TREM1
Clonality: Monoclonal
Clone: 193015
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR