Anti-TREM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 193015]

Supplier: R & D Systems
FAB1278A
102880-308EA 577.02 USD
102880-308
Anti-TREM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 193015]
Antibodies
Type: Primary
Antigen: TREM1
Clonality: Monoclonal
Clone: 193015
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR