Anti-JAG1 Mouse Monoclonal Antibody (Fluorescein) [clone: 188331]

Supplier: R & D Systems
FAB1277F
220017-028EA 454.09 USD
220017-028
Anti-JAG1 Mouse Monoclonal Antibody (Fluorescein) [clone: 188331]
Antibodies
Type: Primary
Antigen: JAG1
Clonality: Monoclonal
Clone: 188331
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR