Anti-JAG1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 188331]

Supplier: R & D Systems
FAB1277A
102880-306EA 608.83 USD
102880-306
Anti-JAG1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 188331]
Antibodies
Type: Primary
Antigen: JAG1
Clonality: Monoclonal
Clone: 188331
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR