Anti-PDGFRA Mouse Monoclonal Antibody (Fluorescein) [clone: PRa292]

Supplier: R & D Systems
FAB1264F
220017-026EA 485.91 USD
220017-026
Anti-PDGFRA Mouse Monoclonal Antibody (Fluorescein) [clone: PRa292]
Antibodies
Type: Primary
Antigen: PDGFRA
Clonality: Monoclonal
Clone: PRa292
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR