Anti-PDGFRB Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: PR7212]

Supplier: R & D Systems
FAB1263C
102880-300EA 584.25 USD
102880-300
Anti-PDGFRB Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: PR7212]
Antibodies
Type: Primary
Antigen: PDGFRB
Clonality: Monoclonal
Clone: PR7212
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR