Anti-TNFRSF4 Goat Polyclonal Antibody (PE (Phycoerythrin))

Supplier: R & D Systems
FAB1256P
102880-296EA 501.82 USD
102880-296
Anti-TNFRSF4 Goat Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: TNFRSF4
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR