Anti-LY6A Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 177228]

Supplier: R & D Systems
FAB1226P
220017-011EA 513.38 USD
220017-011
Anti-LY6A Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 177228]
Antibodies
Type: Primary
Antigen: LY6A
Clonality: Monoclonal
Clone: 177228
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR