Anti-KIR3DL1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: DX9]

Supplier: R & D Systems
FAB1225A-025 FAB1225A-100
102881-142EA 608.83 USD
102881-142 102881-140
Anti-KIR3DL1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: DX9]
Antibodies
Type: Primary
Antigen: KIR3DL1
Clonality: Monoclonal
Clone: DX9
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR