Anti-PDCD1LG2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 176611]

Supplier: R & D Systems
FAB1224A
220017-003EA 577.02 USD
220017-003
Anti-PDCD1LG2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 176611]
Antibodies
Type: Primary
Antigen: PDCD1LG2
Clonality: Monoclonal
Clone: 176611
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR