Anti-ITGAV Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: P2W7]

Supplier: R & D Systems
FAB1219P
220017-002EA 485.91 USD
220017-002
Anti-ITGAV Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: P2W7]
Antibodies
Type: Primary
Antigen: ITGAV
Clonality: Monoclonal
Clone: P2W7
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR