Anti-PTPRC Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RA3-6B2]

Supplier: R & D Systems
FAB1217P
220017-001EA 454.09 USD
220017-001
Anti-PTPRC Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RA3-6B2]
Antibodies
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: RA3-6B2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR