Anti-IL18RAP Rat Monoclonal Antibody (Fluorescein) [clone: 112614]

Supplier: R & D Systems
FAB1216F-100 FAB1216F-025
102881-116EA 193.78 USD
102881-116 102881-118
Anti-IL18RAP Rat Monoclonal Antibody (Fluorescein) [clone: 112614]
Antibodies
Type: Primary
Antigen: IL18RAP
Clonality: Monoclonal
Clone: 112614
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR