Anti-IL18RAP Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 112614]

Supplier: R & D Systems
FAB1216A-025 FAB1216A-100
102881-112EA 274.77 USD
102881-112 102881-114
Anti-IL18RAP Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 112614]
Antibodies
Type: Primary
Antigen: IL18RAP
Clonality: Monoclonal
Clone: 112614
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR