Anti-IL17RB Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 170220]

Supplier: R & D Systems
FAB1207C-025 FAB1207C-100
102881-104EA 300.8 USD
102881-104 102881-106
Anti-IL17RB Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 170220]
Antibodies
Type: Primary
Antigen: IL17RB
Clonality: Monoclonal
Clone: 170220
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR