Anti-LGALS3 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 202213]

Supplier: R & D Systems
FAB1197P
102881-098EA 501.82 USD
102881-098
Anti-LGALS3 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 202213]
Antibodies
Type: Primary
Antigen: LGALS3
Clonality: Monoclonal
Clone: 202213
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR