Anti-IL18RAP Mouse Monoclonal Antibody (Fluorescein) [clone: 132029]

Supplier: R & D Systems
FAB118F-100 FAB118F-025
102881-088EA 152 USD
102881-088 102881-090
Anti-IL18RAP Mouse Monoclonal Antibody (Fluorescein) [clone: 132029]
Antibodies
Type: Primary
Antigen: IL18RAP
Clonality: Monoclonal
Clone: 132029
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR