Anti-JAM3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 208212]

Supplier: R & D Systems
FAB11891P
220016-989EA 527.85 USD
220016-989
Anti-JAM3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 208212]
Antibodies
Type: Primary
Antigen: JAM3
Clonality: Monoclonal
Clone: 208212
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR