Anti-TREM1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 174031]

Supplier: R & D Systems
FAB1187P
220016-987EA 533.63 USD
220016-987
Anti-TREM1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 174031]
Antibodies
Type: Primary
Antigen: TREM1
Clonality: Monoclonal
Clone: 174031
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR