Anti-CD300A Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 172224]

Supplier: R & D Systems
FAB1186P
220016-985EA 454.09 USD
220016-985
Anti-CD300A Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 172224]
Antibodies
Type: Primary
Antigen: CD300A
Clonality: Monoclonal
Clone: 172224
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR