Anti-CD300A Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 172224]

Supplier: R & D Systems
FAB1186A
220016-983EA 519.17 USD
220016-983
Anti-CD300A Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 172224]
Antibodies
Type: Primary
Antigen: CD300A
Clonality: Monoclonal
Clone: 172224
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR