Anti-IL20RA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 173714]

Supplier: R & D Systems
FAB11762P
220016-979EA 543.75 USD
220016-979
Anti-IL20RA Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 173714]
Antibodies
Type: Primary
Antigen: IL20RA
Clonality: Monoclonal
Clone: 173714
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR