Anti-ALCAM Goat Polyclonal Antibody (PE (Phycoerythrin))

Supplier: R & D Systems
FAB1172P
220016-977EA 513.38 USD
220016-977
Anti-ALCAM Goat Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: ALCAM
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR