Anti-ALCAM Goat Polyclonal Antibody (Alexa Fluor® 700)

Supplier: R & D Systems
FAB1172N
102881-080EA 485.91 USD
102881-080
Anti-ALCAM Goat Polyclonal Antibody (Alexa Fluor® 700)
Antibodies
Type: Primary
Antigen: ALCAM
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 700
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR