Anti-ALCAM Goat Polyclonal Antibody (Fluorescein)

Supplier: R & D Systems
FAB1172F
220016-976EA 349.97 USD
220016-976
Anti-ALCAM Goat Polyclonal Antibody (Fluorescein)
Antibodies
Type: Primary
Antigen: ALCAM
Clonality: Polyclonal
Clone:
Conjugation: Fluorescein
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR