Anti-CD40L Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 208109]

Supplier: R & D Systems
FAB1163P
220016-971EA 420.83 USD
220016-971
Anti-CD40L Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 208109]
Antibodies
Type: Primary
Antigen: CD40L
Clonality: Monoclonal
Clone: 208109
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR