Anti-CD5 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 53-7.3]

Supplier: R & D Systems
FAB115P
220016-968EA 268.98 USD
220016-968
Anti-CD5 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 53-7.3]
Antibodies
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 53-7.3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR