Anti-H60A Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 205326]

Supplier: R & D Systems
FAB1155P
220016-966EA 513.38 USD
220016-966
Anti-H60A Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 205326]
Antibodies
Type: Primary
Antigen Symbol: H60A
Antigen Name: histocompatibility 60a
Antigen Synonyms: H60
Clonality: Monoclonal
Clone: 205326
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR