Anti-PTPRC Rat Monoclonal Antibody (Fluorescein) [clone: 30-F11]

Supplier: R & D Systems
FAB114F
220016-963EA 160.52 USD
220016-963
Anti-PTPRC Rat Monoclonal Antibody (Fluorescein) [clone: 30-F11]
Antibodies
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: 30-F11
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR